Assay Details
| Gene Symbol: RGD1564541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107950.1 | NP_001101420.1 | 5 - 6 | 681 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1564541
Genomic Map