Assay Details
| Gene Symbol: Tbc1d22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006242171.2 | XP_006242233.1 | 5 - 6 | 2518 | 63 | |
| XM_008765824.1 | XP_008764046.1 | 5 - 6 | 2516 | 63 | ||
More Information
Set Membership: |
|

Tbc1d22a
Genomic Map