Assay Details
| Gene Symbol: Xkr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012042.1 | NP_001012042.1 | 3 - 4 | 980 | 65 | |
| GenBank mRNA | AY534260.1 | - | 3 - 4 | 980 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P066256 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P066251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162783 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P066251 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P066256 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P066251 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P066256 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P066251 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P066256 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P496832 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68571 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10891 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82924 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38127 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74176 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82307 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 87753 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41568 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8730 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36664 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59768 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104687 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100226 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57779 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67061 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26760 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43162 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82968 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44191 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10781011 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781009 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781008 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781010 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781006 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10781005 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500004877.rn.1 | TC1500000697.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500004881.rn.1 | TC1500000697.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500004879.rn.1 | TC1500000697.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500004880.rn.1 | TC1500000697.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10781005 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21617076 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376928 | - | - |

Xkr6
Genomic Map