Assay Details
| Gene Symbol: Ptbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005555.1 | NP_001005555.1 | 2 - 3 | 123 | 64 | |
| XM_006233236.2 | XP_006233298.1 | 2 - 3 | 657 | 64 | ||
| XM_006233237.2 | XP_006233299.1 | 2 - 3 | 656 | 64 | ||
| XM_006233238.2 | XP_006233300.1 | 2 - 3 | 656 | 64 | ||
| GenBank mRNA | AJ010585.1 | - | 2 - 3 | 92 | 64 | |
| BC082076.1 | - | 2 - 3 | 123 | 7133221 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptbp2
Genomic Map