Assay Details
| Gene Symbol: Mcm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107366.1 | NP_001100836.1 | 18 - 19 | 2672 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P23155 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P262720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143561 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P23155 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P262720 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P23155 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P262720 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P262720 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P23155 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P262720 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67310 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10796359 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796365 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796360 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796358 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796354 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796362 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796350 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796348 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796357 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796351 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796364 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796352 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796363 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796361 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796349 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796353 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796356 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796355 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10796347 | - | - |
| Rat Genome 230 2.0 Array | 1392061_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381298_at | - | - |
| Rat Genome 230 2.0 Array | 1381298_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392061_at | - | - |
| Affymetrix HT Rat Focus Array | 1392061_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392061_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700006918.rn.1 | TC1700001002.rn.1 | RTA TC |
| Rat Expression Set 230 | 1381298_at | - | - |
| Rat Expression Set 230 | 1392061_at | - | - |
| Rat Expression Set 230 | 1381298_PM_at | - | - |
| Rat Expression Set 230 | 1392061_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796347 | - | - |
| Rat Genome U34 Set | rc_AI103253_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21592544 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352298 | - | - |

Mcm10
Genomic Map