Assay Details
| Gene Symbol: Ptrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749449.3 | XP_003749497.2 | 3 - 4 | 516 | 67 | |
| GenBank mRNA | FQ220792.1 | - | 3 - 4 | 401 | 67 | |
More Information
Set Membership: |
|

Ptrh1
Genomic Map