Assay Details
| Gene Symbol: Tmco5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762110.1 | XP_008760332.1 | 7 - 8 | 565 | 85 | |
| XM_575205.5 | XP_575205.2 | 7 - 8 | 509 | 85 | ||
More Information
Set Membership: |
|

Tmco5b
Genomic Map