Assay Details
| Gene Symbol: Dvl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107081.2 | NP_001100551.1 | 7 - 8 | 614 | 87 | |
| XM_006248564.2 | XP_006248626.1 | 7 - 8 | 845 | 87 | ||
| XM_006248565.2 | XP_006248627.1 | 7 - 8 | 770 | 87 | ||
| XM_006248566.2 | XP_006248628.1 | 7 - 8 | 770 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl3
Genomic Map