Assay Details
| Gene Symbol: LOC690911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271495.1 | NP_001258424.1 | 11 - 12 | 9852 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC690911
Genomic Map