Assay Details
| Gene Symbol: LOC690911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271495.1 | NP_001258424.1 | 1 - 2 | 399 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690911| Gene Symbol: LOC690911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271495.1 | NP_001258424.1 | 1 - 2 | 399 | 59 | |
Set Membership: |
|