Assay Details
| Gene Symbol: Timm21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008343.1 | NP_001008344.1 | 4 - 5 | 741 | 92 | |
| GenBank mRNA | BC085823.1 | - | 4 - 5 | 741 | 7126835 | 92 |
More Information
Updated Mapping Information
Set Membership: |
|

Timm21
Genomic Map