Assay Details
| Gene Symbol: U2af2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748763.3 | XP_003748811.1 | 7 - 8 | 840 | 70 | |
| XM_006228320.2 | XP_006228382.1 | 7 - 8 | 840 | 70 | ||
More Information
Set Membership: |
|

U2af2
Genomic Map