Assay Details
| Gene Symbol: Brsk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127337.1 | NP_001120809.1 | 17 - 18 | 2092 | 66 | |
| XM_006228283.1 | XP_006228345.1 | 16 - 17 | 1957 | 66 | ||
| GenBank mRNA | AB365521.1 | - | 17 - 18 | 2092 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brsk1
Genomic Map