Assay Details
| Gene Symbol: Cnppd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199112.1 | NP_954543.1 | 6 - 7 | 822 | 73 | |
| XM_006245226.2 | XP_006245288.1 | 6 - 7 | 856 | 73 | ||
| GenBank mRNA | BC061750.1 | - | 6 - 7 | 822 | 5599704 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|

Cnppd1
Genomic Map