Assay Details
| Gene Symbol: Igfbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012817.1 | NP_036949.1 | 2 - 3 | 1115 | 66 | |
| GenBank mRNA | BC087030.1 | - | 1317 | 66 | ||
| M62781.1 | - | 2 - 3 | 1115 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igfbp5
Genomic Map