Assay Details
| Gene Symbol: LOC501180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024364.1 | NP_001019535.1 | 1 | 1354 | 72 | |
| GenBank mRNA | BC085806.1 | - | 1 | 1354 | 7112277 | 72 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC501180
Genomic Map