Assay Details
| Gene Symbol: Pde6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108806.1 | NP_001102276.1 | 1 - 2 | 175 | 61 | |
| XM_006245413.2 | XP_006245475.1 | 1 - 2 | 223 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108806.1 transcripts.
Set Membership: |
|

Pde6d
Genomic Map