Assay Details
| Gene Symbol: Pde6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108806.1 | NP_001102276.1 | 2 - 3 | 274 | 92 | |
| XM_006245413.2 | XP_006245475.1 | 2 - 3 | 322 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde6d
Genomic Map