Assay Details
| Gene Symbol: Stk11ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106922.2 | NP_001100392.2 | 1 - 2 | 307 | 54 | |
| XM_006245187.2 | XP_006245249.1 | 1 - 2 | 668 | 54 | ||
| XM_006245188.2 | XP_006245250.1 | 1 - 2 | 668 | 54 | ||
| GenBank mRNA | BC168233.1 | - | 1 - 2 | 307 | 7442632 | 54 |
More Information
Updated Mapping Information
Set Membership: |
|

Stk11ip
Genomic Map