Assay Details
Gene Symbol: Slc16a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108229.1 | NP_001101699.1 | 3 - 4 | 782 | 84 | |
XM_006245388.2 | XP_006245450.1 | 3 - 4 | 1011 | 84 | ||
XM_006245389.2 | XP_006245451.1 | 3 - 4 | 1158 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
