Assay Details
| Gene Symbol: Scg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245148.2 | XP_006245210.1 | 2 - 3 | 1173 | 82 | |
| GenBank mRNA | BC061549.1 | - | 2 - 3 | 1025 | 6919444 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|

Scg2
Genomic Map