Assay Details
| Gene Symbol: LOC100366216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750725.3 | XP_003750773.1 | 5 - 6 | 1668 | 65 | |
| XM_006245342.2 | XP_006245404.1 | 6 - 7 | 2329 | 65 | ||
More Information
Set Membership: |
|

LOC100366216
Genomic Map