Assay Details
| Gene Symbol: LOC287004 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110636.1 | 1 - 2 | 237 | 100 | |||
| NR_110638.1 | 1 - 2 | 237 | 100 | ||||
| NR_110640.1 | 1 - 2 | 237 | 100 | ||||
| GenBank mRNA | AY035343.1 | - | LIT1742 PINC |
1 - 2 | 234 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein unc-80 homolog | LOC103693241 | -54.4kb | Sense | Chr. 9 - 72663790 - 72665685 on NCBI Build Rnor_6.0 | |
| Downstream | tensin 1 | Tns1 | Tns | 494.4kb | Sense | Chr. 9 - 81191792 - 81401027 on NCBI Build Rnor_6.0 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC287004
Genomic Map