Assay Details
| Gene Symbol: LOC100911572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750719.3 | XP_003750767.2 | 3 - 4 | 773 | 70 | |
| Gene Symbol: Col4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135759.1 | NP_001129231.1 | 44 - 45 | 3958 | 70 | |
| XM_006245259.2 | XP_006245321.1 | 44 - 45 | 4181 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911572
Genomic Map