Assay Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100760.1 | NP_001094230.1 | 6 - 7 | 842 | 79 | |
| XM_006245244.2 | XP_006245306.1 | 6 - 7 | 864 | 79 | ||
| GenBank mRNA | BC104673.1 | - | 6 - 7 | 652 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam134a
Genomic Map