Assay Details
| Gene Symbol: Alpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022665.3 | NP_073156.3 | 8 - 9 | 1039 | 91 | |
| XM_008767194.1 | XP_008765416.1 | 9 - 10 | 1074 | 91 | ||
| GenBank mRNA | X17611.1 | - | 8 - 9 | 1038 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alpi
Genomic Map