Assay Details
| Gene Symbol: Efhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109310.1 | NP_001102780.1 | 2 - 3 | 544 | 62 | |
| XM_006245468.2 | XP_006245530.1 | 2 - 3 | 580 | 62 | ||
| XM_008767292.1 | XP_008765514.1 | 2 - 3 | 580 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efhd1
Genomic Map