Assay Details
| Gene Symbol: Grb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109093.1 | NP_001102563.1 | 11 - 12 | 1361 | 62 | |
| XM_006251508.2 | XP_006251570.1 | 9 - 10 | 1242 | 62 | ||
| XM_008770347.1 | XP_008768569.1 | 10 - 11 | 1407 | 62 | ||
| XM_008770348.1 | XP_008768570.1 | 9 - 10 | 1242 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
insulin receptor signaling pathway
positive regulation of signal transduction
negative regulation of Wnt signaling pathway
positive regulation of vascular endothelial growth factor receptor signaling pathway
response to insulin
negative regulation of phosphorylation
positive regulation of phosphorylation
negative regulation of glycogen biosynthetic process
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
molecular_function
SH3/SH2 adaptor activity
insulin receptor binding
biological_process
insulin receptor signaling pathway
positive regulation of signal transduction
negative regulation of Wnt signaling pathway
positive regulation of vascular endothelial growth factor receptor signaling pathway
response to insulin
negative regulation of phosphorylation
positive regulation of phosphorylation
negative regulation of glycogen biosynthetic process
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
molecular_function
SH3/SH2 adaptor activity
insulin receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P943285 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P074904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113815 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074904 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P943285 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P943285 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074904 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P074904 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P943285 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P473908 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P417077 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P943285 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78366 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97251 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71400 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11619 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10778513 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778516 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778510 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778515 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778509 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778518 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778521 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778512 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778514 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778507 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778517 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778508 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778519 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778511 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778520 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10778505 | - | - |
| Rat Genome 230 2.0 Array | 1371517_at | - | - |
| Rat Genome 230 2.0 Array | 1397211_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1394610_at | - | - |
| Rat Genome 230 2.0 Array | 1394610_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397211_at | - | - |
| Rat Genome 230 2.0 Array | 1371517_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394610_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397211_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371517_at | - | - |
| Affymetrix HT Rat Focus Array | 1397211_at | - | - |
| Affymetrix HT Rat Focus Array | 1394610_at | - | - |
| Affymetrix HT Rat Focus Array | 1371517_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400016843.rn.1 | TC1400002248.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371517_PM_at | - | - |
| Rat Expression Set 230 | 1371517_at | - | - |
| Rat Expression Set 230 | 1397211_at | - | - |
| Rat Expression Set 230 | 1397211_PM_at | - | - |
| Rat Expression Set 230 | 1394610_at | - | - |
| Rat Expression Set 230 | 1394610_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778505 | - | - |
| Rat Genome U34 Set | rc_AA800686_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21635615 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370918 | - | - |

Grb10
Genomic Map