Assay Details
| Gene Symbol: Igfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012588.2 | NP_036720.2 | 3 - 4 | 860 | 78 | |
| GenBank mRNA | BF281457.1 | - | 2 - 3 | 323 | 78 | |
| M31837.1 | - | 3 - 4 | 813 | 78 | ||
| M33300.1 | - | 3 - 4 | 790 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igfbp3
Genomic Map