Assay Details
| Gene Symbol: Ttc39c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077231.1 | NP_001070699.1 | 8 - 9 | 1097 | 62 | |
| XM_006254423.2 | XP_006254485.1 | 8 - 9 | 1855 | 62 | ||
| XM_008771954.1 | XP_008770176.1 | 7 - 8 | 953 | 62 | ||
| GenBank mRNA | BC105623.1 | - | 8 - 9 | 1097 | 7455888 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc39c
Genomic Map