Assay Details
| Gene Symbol: Aqp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142366.2 | NP_001135838.1 | 3 - 4 | 679 | 85 | |
| NM_012825.4 | NP_036957.1 | 3 - 4 | 671 | 85 | ||
| GenBank mRNA | U14007.1 | - | 3 - 4 | 605 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aqp4
Genomic Map