Assay Details
Gene Symbol: LOC689991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767349.1 | XP_008765571.1 | 10 - 11 | 1670 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC689991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767349.1 | XP_008765571.1 | 10 - 11 | 1670 | 109 |
Set Membership: |
![]() ![]() ![]() |