Assay Details
Gene Symbol: LOC689991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767349.1 | XP_008765571.1 | 1 - 2 | 213 | 135 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008767349.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
