Assay Details
| Gene Symbol: Arhgef3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106061.1 | NP_001099531.1 | 2 - 3 | 322 | 57 | |
| XM_006252574.2 | XP_006252636.1 | 5 - 6 | 454 | 57 | ||
| XM_006252575.2 | XP_006252637.1 | 5 - 6 | 652 | 57 | ||
| XM_006252576.2 | XP_006252638.1 | 3 - 4 | 818 | 57 | ||
| XM_008770982.1 | XP_008769204.1 | 5 - 6 | 519 | 57 | ||
| XM_008770983.1 | XP_008769205.1 | 5 - 6 | 361 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef3
Genomic Map