Assay Details
| Gene Symbol: Slc9a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773665.1 | XP_008771887.1 | 2 - 3 | 288 | 62 | |
| XM_217630.8 | XP_217630.3 | 1 - 2 | 328 | 62 | ||
More Information
Set Membership: |
|

Slc9a6
Genomic Map