Assay Details
| Gene Symbol: Cd40lg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053353.1 | NP_445805.1 | 2 - 3 | 288 | 102 | |
| GenBank mRNA | AF116582.1 | - | 2 - 3 | 288 | 102 | |
| EF066490.1 | - | 2 - 3 | 288 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd40lg
Genomic Map