Assay Details
| Gene Symbol: Hars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025414.1 | NP_001020585.1 | 4 - 5 | 415 | 83 | |
| GenBank mRNA | BC097969.1 | - | 4 - 5 | 415 | 7442005 | 83 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hars
Genomic Map