Assay Details
| Gene Symbol: Nadk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044252.1 | NP_001037717.1 | 5 - 6 | 508 | 92 | |
| XM_006232014.2 | XP_006232076.1 | 4 - 5 | 736 | 92 | ||
| XM_006232015.2 | XP_006232077.1 | 4 - 5 | 735 | 92 | ||
| XM_006232016.2 | XP_006232078.1 | 5 - 6 | 522 | 92 | ||
| GenBank mRNA | BC105901.1 | - | 1 - 2 | 92 | 7375971 | 92 |
| DQ504300.1 | - | 5 - 6 | 508 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nadk2
Genomic Map