Assay Details
| Gene Symbol: Rpl37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031106.1 | NP_112368.1 | 2 - 3 | 178 | 61 | |
| GenBank mRNA | BC059132.1 | - | 2 - 3 | 175 | 6919306 | 61 |
| BC069173.1 | - | 2 - 3 | 160 | 7103990 | 61 | |
| FQ210660.1 | - | 2 - 3 | 181 | 61 | ||
| FQ210874.1 | - | 2 - 3 | 181 | 61 | ||
| FQ217102.1 | - | 2 - 3 | 181 | 61 | ||
| FQ217508.1 | - | 2 - 3 | 171 | 61 | ||
| FQ220738.1 | - | 2 - 3 | 181 | 61 | ||
| FQ221496.1 | - | 2 - 3 | 171 | 61 | ||
| FQ221700.1 | - | 2 - 3 | 171 | 61 | ||
| FQ222983.1 | - | 2 - 3 | 181 | 61 | ||
| FQ223019.1 | - | 2 - 3 | 181 | 61 | ||
| FQ223296.1 | - | 2 - 3 | 186 | 61 | ||
| FQ223471.1 | - | 2 - 3 | 171 | 61 | ||
| S79981.1 | - | 2 - 3 | 178 | 61 | ||
| X66369.1 | - | 2 - 3 | 178 | 61 | ||
| Gene Symbol: LOC100360841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002730037.4 | XP_002730083.1 | 1 | 174 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl37
Genomic Map