Assay Details
| Gene Symbol: Msmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080886.1 | NP_543162.1 | 3 - 4 | 485 | 104 | |
| GenBank mRNA | BC063155.1 | - | 3 - 4 | 472 | 6918081 | 104 |
| D50559.1 | - | 3 - 4 | 485 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msmo1
Genomic Map