Assay Details
| Gene Symbol: Tgfbr3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001067872.5 | XP_001067872.3 | 4 - 5 | 946 | 109 | |
| XM_006248789.2 | XP_006248851.1 | 3 - 4 | 1152 | 109 | ||
| XM_006248790.2 | XP_006248852.1 | 3 - 4 | 1152 | 109 | ||
| XM_008769017.1 | XP_008767239.1 | 3 - 4 | 1152 | 109 | ||
More Information
Set Membership: |
|

Tgfbr3l
Genomic Map