Assay Details
| Gene Symbol: Pcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768953.1 | XP_008767175.1 | 1 - 2 | 92 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768953.1 transcripts.
Set Membership: |
|

Pcp2
Genomic Map