Assay Details
| Gene Symbol: Rab6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108775.1 | NP_001102245.1 | 4 - 5 | 336 | 60 | |
| XM_006243669.2 | XP_006243731.1 | 5 - 6 | 640 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab6b
Genomic Map