Assay Details
| Gene Symbol: Ss18l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750606.3 | XP_003750654.1 | 1 - 2 | 150 | 66 | |
| GenBank mRNA | FQ229074.1 | - | 1 - 2 | 96 | 66 | |
More Information
Set Membership: |
|

Ss18l2
Genomic Map