Assay Details
| Gene Symbol: Onecut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243404.2 | XP_006243466.1 | 1 - 2 | 1519 | 75 | |
| GenBank mRNA | Y14933.1 | - | 1 - 2 | 1140 | 75 | |
More Information
Set Membership: |
|

Onecut1
Genomic Map