Assay Details
| Gene Symbol: Onecut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243404.2 | XP_006243466.1 | 2 - 3 | 1588 | 84 | |
| GenBank mRNA | Y14933.1 | - | 2 - 3 | 1209 | 84 | |
More Information
Set Membership: |
|

Onecut1
Genomic Map