Assay Details
| Gene Symbol: Mzt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105860.1 | NP_001099330.1 | 1 - 2 | 640 | 59 | |
| XM_006248729.1 | XP_006248791.1 | 2 - 3 | 272 | 59 | ||
| XM_006248730.2 | XP_006248792.1 | 2 - 3 | 712 | 59 | ||
| XM_006248731.2 | XP_006248793.1 | 2 - 3 | 706 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mzt2b
Genomic Map