Assay Details
| Gene Symbol: Il1rap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167840.1 | NP_001161312.1 | 9 - 10 | 1240 | 65 | |
| NM_012968.1 | NP_037100.1 | 8 - 9 | 1159 | 65 | ||
| XM_006248504.2 | XP_006248566.1 | 8 - 9 | 1228 | 65 | ||
| GenBank mRNA | GU123169.1 | - | 9 - 10 | 1240 | 65 | |
| U48592.1 | - | 8 - 9 | 1159 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P156612 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P134370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79507 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P134370 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156612 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P134370 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156612 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P134370 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P156612 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1009757 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701897104 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7090 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28515 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78346 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35272 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15983 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50137 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41794 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86012 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23861 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80711 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12746 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33781 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701897104 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10755020 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755024 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755021 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755019 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755015 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755018 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755017 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755016 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755025 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755023 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755026 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755022 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10755013 | - | - |
| Rat Genome 230 2.0 Array | 1393563_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387507_at | - | - |
| Rat Genome 230 2.0 Array | 1387507_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393563_at | - | - |
| Affymetrix HT Rat Focus Array | 1387507_at | - | - |
| Affymetrix HT Rat Focus Array | 1387507_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393563_at | - | - |
| Affymetrix HT Rat Focus Array | 1393563_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100011703.rn.1 | TC1100001837.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387507_PM_at | - | - |
| Rat Expression Set 230 | 1387507_at | - | - |
| Rat Expression Set 230 | 1393563_at | - | - |
| Rat Expression Set 230 | 1393563_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755013 | - | - |
| Rat Genome U34 Set | U48592_g_at | - | - |
| Rat Genome U34 Set | U48592_at | - | - |
| Rat Genome U34 Set | rc_AA819412_at | - | - |
| Rat Genome U34 Set | U48592_at | - | - |
| Rat Genome U34 Set | U48592_g_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355895 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62377 | - | - |

Il1rap
Genomic Map