Assay Details
| Gene Symbol: Tango2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108323.1 | NP_001101793.1 | 2 - 3 | 187 | 71 | |
| XM_006248628.2 | XP_006248690.1 | 2 - 3 | 312 | 71 | ||
| XM_006248629.2 | XP_006248691.1 | 2 - 3 | 182 | 71 | ||
| XM_006248630.2 | XP_006248692.1 | 2 - 3 | 207 | 71 | ||
| XM_006248632.2 | XP_006248694.1 | 2 - 3 | 312 | 71 | ||
| XM_006248633.2 | XP_006248695.1 | 2 - 3 | 311 | 71 | ||
| XM_006248634.2 | XP_006248696.1 | 2 - 3 | 313 | 71 | ||
| XM_008768888.1 | XP_008767110.1 | 2 - 3 | 185 | 71 | ||
| XM_008768889.1 | XP_008767111.1 | 2 - 3 | 312 | 71 | ||
| GenBank mRNA | FQ217948.1 | - | 2 - 3 | 314 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tango2
Genomic Map