Assay Details
| Gene Symbol: Dgcr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107080.2 | NP_001100550.1 | 3 - 4 | 384 | 77 | |
| GenBank mRNA | BC158828.1 | - | 3 - 4 | 384 | 7378165 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Dgcr6
Genomic Map